siRNA Details
| Gene Symbol: Mustn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181368.3 | NP_852033.1 | 2 | 373 | ||
| GenBank mRNA | AY254906.2 | - | 2 | 373 | ||
| BC158570.1 | - | 2 | 388 | 9092791 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mustn1| Gene Symbol: Mustn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181368.3 | NP_852033.1 | 2 | 373 | ||
| GenBank mRNA | AY254906.2 | - | 2 | 373 | ||
| BC158570.1 | - | 2 | 388 | 9092791 | ||