siRNA Details
| Gene Symbol: Zfp488 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006252711.2 | XP_006252773.1 | 2 | 798 | ||
| XM_006252712.2 | XP_006252774.1 | 2 | 732 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp488| Gene Symbol: Zfp488 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006252711.2 | XP_006252773.1 | 2 | 798 | ||
| XM_006252712.2 | XP_006252774.1 | 2 | 732 | |||