siRNA Details
| Gene Symbol: LOC685463 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_001063889.4 | XP_001063889.2 | 355 | |||
| XM_008771169.1 | XP_008769391.1 | 481 | ||||
| Gene Symbol: LOC100911649 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003751563.3 | XP_003751611.1 | 2 | 689 | ||
| XM_008771171.1 | XP_008769393.1 | 1 | 815 | |||
| Gene Symbol: RGD1562641 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_224663.7 | XP_224663.5 | 2 | 1613 | ||

LOC685463
Genomic Map