siRNA Details
| Gene Symbol: Mrpl34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001006965.1 | NP_001006966.1 | 2 | 464 | ||
| XM_006252859.2 | XP_006252921.1 | 3 | 1339 | |||
| GenBank mRNA | BC084718.1 | - | 2 | 464 | 7316058 | |
| FQ217836.1 | - | 2 | 457 | |||

Mrpl34
Genomic Map