siRNA Details
| Gene Symbol: Gtpbp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011919.1 | NP_001011919.1 | 3 | 391 | ||
| GenBank mRNA | BC087083.1 | - | 3 | 391 | 7189218 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gtpbp3| Gene Symbol: Gtpbp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011919.1 | NP_001011919.1 | 3 | 391 | ||
| GenBank mRNA | BC087083.1 | - | 3 | 391 | 7189218 | |