siRNA Details
| Gene Symbol: Mvb12a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199375.1 | NP_955407.1 | 4 | 416 | ||
| XM_006252860.2 | XP_006252922.1 | 4 | 451 | |||
| GenBank mRNA | BC061799.1 | - | 4 | 416 | 5598467 | |
| FQ231372.1 | - | 4 | 453 | |||
| FQ233380.1 | - | 4 | 453 | |||
| FQ234388.1 | - | 4 | 453 | |||

Mvb12a
Genomic Map