siRNA Details
| Gene Symbol: Ifi30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001030026.1 | NP_001025197.1 | 2, 3 | 438 | ||
| GenBank mRNA | BC088256.1 | - | 2, 3 | 302 | ||
| BC099774.1 | - | 2, 3 | 438 | 7932222 | ||
| FQ225096.1 | - | 2, 3 | 324 | |||
| FQ229307.1 | - | 2, 3 | 476 | |||
| FQ230297.1 | - | 2, 3 | 324 | |||
| FQ232607.1 | - | 2, 3 | 324 | |||
| FQ233058.1 | - | 2, 3 | 324 | |||
| FQ233063.1 | - | 2, 3 | 356 | |||
| FQ233214.1 | - | 2, 3 | 324 | |||
| FQ234315.1 | - | 2, 3 | 324 | |||

Ifi30
Genomic Map