siRNA Details
| Gene Symbol: Mpv17l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106072.2 | NP_001099542.1 | 2 | 294 | ||
| XM_008771087.1 | XP_008769309.1 | 2 | 299 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mpv17l2| Gene Symbol: Mpv17l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106072.2 | NP_001099542.1 | 2 | 294 | ||
| XM_008771087.1 | XP_008769309.1 | 2 | 299 | |||