siRNA Details
| Gene Symbol: Pgpep1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_201988.2 | NP_973717.1 | 3 | 147 | ||
| GenBank mRNA | AB098134.1 | - | 4 | 142 | ||
| BC098645.1 | - | 3 | 147 | 7108516 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pgpep1| Gene Symbol: Pgpep1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_201988.2 | NP_973717.1 | 3 | 147 | ||
| GenBank mRNA | AB098134.1 | - | 4 | 142 | ||
| BC098645.1 | - | 3 | 147 | 7108516 | ||