siRNA Details
| Gene Symbol: Isyna1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013880.2 | NP_001013902.2 | 6 | 737 | ||
| GenBank mRNA | BC079011.1 | - | 4 | 741 | ||
| BP490872.1 | - | 2 | 198 | |||
| FQ229915.1 | - | 1 | 33 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Isyna1| Gene Symbol: Isyna1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013880.2 | NP_001013902.2 | 6 | 737 | ||
| GenBank mRNA | BC079011.1 | - | 4 | 741 | ||
| BP490872.1 | - | 2 | 198 | |||
| FQ229915.1 | - | 1 | 33 | |||