siRNA Details
| Gene Symbol: TULP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AB593123.1 | - | 8 | 822 | ||
| AB593124.1 | - | 8 | 808 | |||
| BC032714.1 | - | 7 | 648 | 5495136 | ||
| BC065261.1 | - | 7 | 631 | 6144003 | ||
| U82468.1 | - | 8 | 814 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TULP1| Gene Symbol: TULP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AB593123.1 | - | 8 | 822 | ||
| AB593124.1 | - | 8 | 808 | |||
| BC032714.1 | - | 7 | 648 | 5495136 | ||
| BC065261.1 | - | 7 | 631 | 6144003 | ||
| U82468.1 | - | 8 | 814 | |||