siRNA Details
| Gene Symbol: Gatad2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013881.2 | NP_001013903.2 | 7, 8 | 1292 | ||
| XM_006252879.2 | XP_006252941.1 | 6, 7 | 972 | |||
| XM_006252881.2 | XP_006252943.1 | 6, 7 | 963 | |||
| XM_006252883.2 | XP_006252945.1 | 6, 7 | 972 | |||
| XM_006252884.2 | XP_006252946.1 | 7, 8 | 1251 | |||
| XM_006252887.2 | XP_006252949.1 | 6, 7 | 784 | |||
| XM_008771089.1 | XP_008769311.1 | 8, 9 | 1341 | |||
| XM_008771090.1 | XP_008769312.1 | 7, 8 | 971 | |||
| GenBank mRNA | BC089902.1 | - | 4, 5 | 382 | 7383529 | |

Gatad2a
Genomic Map