siRNA Details
| Gene Symbol: Ints10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134416.1 | NP_001127888.1 | 8 | 933 | ||
| XM_008771092.1 | XP_008769314.1 | 8 | 1213 | |||
| GenBank mRNA | BC166856.1 | - | 8 | 906 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ints10| Gene Symbol: Ints10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134416.1 | NP_001127888.1 | 8 | 933 | ||
| XM_008771092.1 | XP_008769314.1 | 8 | 1213 | |||
| GenBank mRNA | BC166856.1 | - | 8 | 906 | ||