siRNA Details
| Gene Symbol: Ints10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134416.1 | NP_001127888.1 | 3 | 302 | ||
| XM_008771092.1 | XP_008769314.1 | 3 | 582 | |||
| GenBank mRNA | BC166856.1 | - | 3 | 275 | ||
| CB807605.1 | - | 3 | 302 | |||

Ints10
Genomic Map