siRNA Details
Gene Symbol: Tktl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106080.1 | NP_001099550.1 | 1 | 945 | ||
XM_006253022.2 | XP_006253084.1 | 1 | 970 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Tktl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106080.1 | NP_001099550.1 | 1 | 945 | ||
XM_006253022.2 | XP_006253084.1 | 1 | 970 |