siRNA Details
| Gene Symbol: Klhl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008771199.1 | XP_008769421.1 | 11 | 1585 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Klhl2| Gene Symbol: Klhl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008771199.1 | XP_008769421.1 | 11 | 1585 | ||