siRNA Details
| Gene Symbol: Cep44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037181.1 | NP_001032258.1 | 8 | 1163 | ||
| XM_006253087.2 | XP_006253149.1 | 7 | 992 | |||
| XM_006253089.2 | XP_006253151.1 | 8 | 1137 | |||
| XM_006253090.2 | XP_006253152.1 | 6 | 851 | |||
| XM_008771223.1 | XP_008769445.1 | 8 | 1422 | |||
| GenBank mRNA | BC107470.1 | - | 8 | 1163 | 7457548 | |
| FQ228732.1 | - | 8 | 1137 | |||

Cep44
Genomic Map