siRNA Details
| Gene Symbol: TULP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001160408.1 | NP_001153880.1 | 2 | 163 | ||
| NM_003324.4 | NP_003315.2 | 2 | 163 | |||
| GenBank mRNA | AF045583.1 | - | 2 | 127 | ||
| AK024246.1 | - | 2 | 163 | |||
| AK293285.1 | - | 2 | 142 | |||
| AK307434.1 | - | 2 | 139 | |||
| BC032587.1 | - | 2 | 136 | 5501868 | ||

TULP3
Genomic Map