siRNA Details
| Gene Symbol: Lrp2bp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017443.1 | NP_001017443.1 | 4 | 465 | ||
| XM_006253141.2 | XP_006253203.1 | 4 | 381 | |||
| XM_006253143.2 | XP_006253205.1 | 5 | 1462 | |||
| GenBank mRNA | BC092575.1 | - | 4 | 465 | 7114719 | |

Lrp2bp
Genomic Map