siRNA Details
| Gene Symbol: F11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001047848.1 | NP_001041313.1 | 9 | 1159 | ||
| GenBank mRNA | AY387072.1 | - | 9 | 1159 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

F11| Gene Symbol: F11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001047848.1 | NP_001041313.1 | 9 | 1159 | ||
| GenBank mRNA | AY387072.1 | - | 9 | 1159 | ||