siRNA Details
| Gene Symbol: Adam26a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001169119.1 | NP_001162590.1 | 329 | |||
| GenBank mRNA | BC085958.1 | - | 329 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Adam26a| Gene Symbol: Adam26a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001169119.1 | NP_001162590.1 | 329 | |||
| GenBank mRNA | BC085958.1 | - | 329 | |||