siRNA Details
Gene Symbol: Vps37a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024867.1 | NP_001020038.1 | 5, 6 | 879 | ||
XM_008771289.1 | XP_008769511.1 | 5, 6 | 941 | |||
GenBank mRNA | BC098069.1 | - | 5, 6 | 879 | 7444656 | |
FQ232149.1 | - | 5, 6 | 933 |
