siRNA Details
| Gene Symbol: Vps37a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024867.1 | NP_001020038.1 | 9, 10 | 1214 | ||
| GenBank mRNA | BC098069.1 | - | 9, 10 | 1214 | 7444656 | |
| FQ232149.1 | - | 9, 10 | 1229 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vps37a| Gene Symbol: Vps37a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024867.1 | NP_001020038.1 | 9, 10 | 1214 | ||
| GenBank mRNA | BC098069.1 | - | 9, 10 | 1214 | 7444656 | |
| FQ232149.1 | - | 9, 10 | 1229 | |||