siRNA Details
| Gene Symbol: Tusc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004212.1 | NP_001004212.1 | 4, 5 | 754 | ||
| XM_008771294.1 | XP_008769516.1 | 4, 5 | 772 | |||
| GenBank mRNA | BC079193.1 | - | 4, 5 | 754 | 7131345 | |
| FQ212588.1 | - | 3, 4 | 350 | |||
| FQ213307.1 | - | 4, 5 | 772 | |||

Tusc3
Genomic Map