siRNA Details
| Gene Symbol: LOC103693972 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008771305.1 | XP_008769527.1 | 9 | 2102 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC103693972| Gene Symbol: LOC103693972 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008771305.1 | XP_008769527.1 | 9 | 2102 | ||