siRNA Details
| Gene Symbol: Erlin2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106088.2 | NP_001099558.1 | 5, 6 | 353 | ||
| XM_008771323.1 | XP_008769545.1 | 5, 6 | 606 | |||
| XM_008771324.1 | XP_008769546.1 | 5, 6 | 548 | |||
| GenBank mRNA | BC168668.1 | - | 5, 6 | 353 | 7934033 | |

Erlin2
Genomic Map