siRNA Details
| Gene Symbol: Ash2l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106089.1 | NP_001099559.1 | 7, 8 | 782 | ||
| XM_006253303.2 | XP_006253365.1 | 7, 8 | 808 | |||
| XM_006253304.2 | XP_006253366.1 | 7, 8 | 729 | |||
| GenBank mRNA | FQ233069.1 | - | 4, 5 | 385 | ||

Ash2l
Genomic Map