siRNA Details
| Gene Symbol: Ash2l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106089.1 | NP_001099559.1 | 1 | 94 | ||
| XM_006253303.2 | XP_006253365.1 | 1 | 120 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ash2l| Gene Symbol: Ash2l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106089.1 | NP_001099559.1 | 1 | 94 | ||
| XM_006253303.2 | XP_006253365.1 | 1 | 120 | |||