siRNA Details
| Gene Symbol: Gins4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001030027.1 | NP_001025198.1 | 4, 5 | 456 | ||
| GenBank mRNA | BC099741.1 | - | 4, 5 | 456 | 5623828 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gins4| Gene Symbol: Gins4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001030027.1 | NP_001025198.1 | 4, 5 | 456 | ||
| GenBank mRNA | BC099741.1 | - | 4, 5 | 456 | 5623828 | |