siRNA Details
| Gene Symbol: Agpat6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001047849.1 | NP_001041314.1 | 11 | 2037 | ||
| GenBank mRNA | BC082092.1 | - | 8 | 883 | ||
| BC161809.1 | - | 10 | 1323 | 8374387 | ||
| DQ480750.1 | - | 11 | 2037 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Agpat6| Gene Symbol: Agpat6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001047849.1 | NP_001041314.1 | 11 | 2037 | ||
| GenBank mRNA | BC082092.1 | - | 8 | 883 | ||
| BC161809.1 | - | 10 | 1323 | 8374387 | ||
| DQ480750.1 | - | 11 | 2037 | |||