siRNA Details
| Gene Symbol: Mrps31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106091.1 | NP_001099561.1 | 4 | 719 | ||
| XM_006253349.2 | XP_006253411.1 | 1940 | ||||
| GenBank mRNA | BC158691.1 | - | 4 | 705 | 9021908 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mrps31| Gene Symbol: Mrps31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106091.1 | NP_001099561.1 | 4 | 719 | ||
| XM_006253349.2 | XP_006253411.1 | 1940 | ||||
| GenBank mRNA | BC158691.1 | - | 4 | 705 | 9021908 | |