siRNA Details
| Gene Symbol: Qars | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007624.1 | NP_001007625.1 | 4 | 756 | ||
| XM_008766477.1 | XP_008764699.1 | 4 | 750 | |||
| GenBank mRNA | BC082002.1 | - | 4 | 756 | 7131336 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Qars| Gene Symbol: Qars | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007624.1 | NP_001007625.1 | 4 | 756 | ||
| XM_008766477.1 | XP_008764699.1 | 4 | 750 | |||
| GenBank mRNA | BC082002.1 | - | 4 | 756 | 7131336 | |