siRNA Details
Gene Symbol: Cdc16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024744.1 | NP_001019915.1 | 17 | 1701 | ||
GenBank mRNA | BC097498.1 | - | 17 | 1701 | 7462333 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Cdc16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024744.1 | NP_001019915.1 | 17 | 1701 | ||
GenBank mRNA | BC097498.1 | - | 17 | 1701 | 7462333 |