siRNA Details
| Gene Symbol: Fastkd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001082574.1 | NP_001076043.1 | 2 | 804 | ||
| XM_006227809.2 | XP_006227871.1 | 2 | 1576 | |||
| XM_006227811.2 | XP_006227873.1 | 2 | 1017 | |||
| XM_008758678.1 | XP_008756900.1 | 1 | 1973 | |||
| XM_008758679.1 | XP_008756901.1 | 2 | 1880 | |||
| GenBank mRNA | BC079475.1 | - | 1 | 562 | ||
| BC161806.1 | - | 1 | 775 | 7111194 | ||

Fastkd3
Genomic Map