siRNA Details
| Gene Symbol: Slc35d2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106098.2 | NP_001099568.2 | 9 | 728 | ||
| XM_006253493.2 | XP_006253555.1 | 9 | 1712 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc35d2| Gene Symbol: Slc35d2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106098.2 | NP_001099568.2 | 9 | 728 | ||
| XM_006253493.2 | XP_006253555.1 | 9 | 1712 | |||