siRNA Details
| Gene Symbol: Ctsr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175581.3 | NP_783171.2 | 5 | 582 | ||
| XM_008771434.1 | XP_008769656.1 | 4 | 503 | |||
| GenBank mRNA | BC097484.1 | - | 5 | 582 | 7459242 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ctsr| Gene Symbol: Ctsr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175581.3 | NP_783171.2 | 5 | 582 | ||
| XM_008771434.1 | XP_008769656.1 | 4 | 503 | |||
| GenBank mRNA | BC097484.1 | - | 5 | 582 | 7459242 | |