siRNA Details
| Gene Symbol: Isca1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181626.3 | NP_853657.1 | 4 | 314 | ||
| GenBank mRNA | AF508158.1 | - | 4 | 280 | ||
| BC070929.1 | - | 4 | 314 | 7096424 | ||
| BC078677.1 | - | 4 | 324 | 7133788 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Isca1| Gene Symbol: Isca1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181626.3 | NP_853657.1 | 4 | 314 | ||
| GenBank mRNA | AF508158.1 | - | 4 | 280 | ||
| BC070929.1 | - | 4 | 314 | 7096424 | ||
| BC078677.1 | - | 4 | 324 | 7133788 | ||