siRNA Details
| Gene Symbol: Agtpbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106100.1 | NP_001099570.1 | 5, 6 | 551 | ||
| XM_008771435.1 | XP_008769657.1 | 5, 6 | 1935 | |||
| XM_008771436.1 | XP_008769658.1 | 5, 6 | 1935 | |||
| XM_008771437.1 | XP_008769659.1 | 3, 4 | 493 | |||
| GenBank mRNA | BC158657.1 | - | 3, 4 | 229 | ||

Agtpbp1
Genomic Map