siRNA Details
| Gene Symbol: Prelid1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009636.1 | NP_001009636.1 | 4 | 588 | ||
| GenBank mRNA | BC088284.1 | - | 4 | 588 | 7323653 | |
| FQ212767.1 | - | 4 | 600 | |||
| FQ226295.1 | - | 4 | 602 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_025596, AK002846, AK012144, AK153455, BC024813, and others.

Prelid1
Genomic Map