siRNA Details
| Gene Symbol: Higd2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106102.1 | NP_001099572.1 | 2 | 530 | ||
| GenBank mRNA | BC166544.1 | - | 2 | 500 | 7301761 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Higd2a| Gene Symbol: Higd2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106102.1 | NP_001099572.1 | 2 | 530 | ||
| GenBank mRNA | BC166544.1 | - | 2 | 500 | 7301761 | |