siRNA Details
| Gene Symbol: Ogn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106103.1 | NP_001099573.1 | 6 | 836 | ||
| XM_006253696.2 | XP_006253758.2 | 7 | 1282 | |||
| XM_006253699.2 | XP_006253761.1 | 7 | 1282 | |||
| XM_008771490.1 | XP_008769712.1 | 7 | 1282 | |||
| XM_008771558.1 | XP_008769780.1 | 7 | 1282 | |||
| GenBank mRNA | FQ213759.1 | - | 5 | 748 | ||
| FQ221052.1 | - | 5 | 748 | |||

Ogn
Genomic Map