siRNA Details
| Gene Symbol: Id4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175582.1 | NP_783172.1 | 1 | 282 | ||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_031166, AK041164, X75018, BC145367.

Id4
Genomic Map