siRNA Details
| Gene Symbol: Fam8a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008771560.1 | XP_008769782.1 | 4 | 1013 | ||
| XM_214446.7 | XP_214446.4 | 5 | 1147 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam8a1| Gene Symbol: Fam8a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008771560.1 | XP_008769782.1 | 4 | 1013 | ||
| XM_214446.7 | XP_214446.4 | 5 | 1147 | |||