siRNA Details
Gene Symbol: Nedd9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011922.1 | NP_001011922.1 | 5 | 1983 | ||
XM_006253786.2 | XP_006253848.1 | 5 | 2146 | |||
XM_008771569.1 | XP_008769791.1 | 5 | 2799 | |||
GenBank mRNA | BC085812.1 | - | 5 | 1983 | 7121465 |
