siRNA Details
| Gene Symbol: RGD1307537 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013885.2 | NP_001013907.1 | 3 | 682 | ||
| GenBank mRNA | BC079269.1 | - | 683 | 7133712 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1307537| Gene Symbol: RGD1307537 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013885.2 | NP_001013907.1 | 3 | 682 | ||
| GenBank mRNA | BC079269.1 | - | 683 | 7133712 | ||