siRNA Details
| Gene Symbol: Dcdc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106110.2 | NP_001099580.2 | 3 | 453 | ||
| GenBank mRNA | CV111304.1 | - | 3 | 161 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dcdc2| Gene Symbol: Dcdc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106110.2 | NP_001099580.2 | 3 | 453 | ||
| GenBank mRNA | CV111304.1 | - | 3 | 161 | ||