siRNA Details
| Gene Symbol: Gmnn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106112.1 | NP_001099582.1 | 4 | 604 | ||
| XM_006253927.2 | XP_006253989.1 | 4 | 389 | |||
| XM_006253928.2 | XP_006253990.1 | 4 | 327 | |||
| GenBank mRNA | FQ219913.1 | - | 1 | 92 | ||

Gmnn
Genomic Map