siRNA Details
| Gene Symbol: Hist1h1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106113.1 | NP_001099583.1 | 1 | 371 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hist1h1a| Gene Symbol: Hist1h1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106113.1 | NP_001099583.1 | 1 | 371 | ||