siRNA Details
| Gene Symbol: Gpr141 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181767.1 | NP_861432.1 | 1 | 359 | ||
| GenBank mRNA | AY288432.1 | - | 1 | 359 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gpr141| Gene Symbol: Gpr141 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181767.1 | NP_861432.1 | 1 | 359 | ||
| GenBank mRNA | AY288432.1 | - | 1 | 359 | ||
Set Membership: |
|