siRNA Details
| Gene Symbol: Stard3nl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008298.1 | NP_001008299.1 | 6, 7 | 668 | ||
| GenBank mRNA | BC086352.1 | - | 6, 7 | 668 | 7303026 | |
| FQ220233.1 | - | 6, 7 | 691 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Stard3nl| Gene Symbol: Stard3nl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008298.1 | NP_001008299.1 | 6, 7 | 668 | ||
| GenBank mRNA | BC086352.1 | - | 6, 7 | 668 | 7303026 | |
| FQ220233.1 | - | 6, 7 | 691 | |||